VWR®, Liquid-in-Glass Refrigerator/Freezer Thermometer
Supplier: VWR Collection
VWRI620-0876EA 36.7 SGD
VWRI620-0876
VWR®, Liquid-in-Glass Refrigerator/Freezer Thermometer
Thermometers Analogue Thermometers
Dual-scale thermometer (°C/°F) with completely enclosed bulb for accurate readings. Easy to read on white-backed glass.
  • Red spirit
  • Rust-proof case
  • Stainless steel hangers
  • Enamelled aluminum scale
W×D: 28×124 mm
W×D: 28×124 mm