VWR®, Digital Thermo-Hygrometer
Supplier: VWR Collection
VWRI620-0915EA 132 SGD
VWRI620-0915
VWR®, Digital Thermo-Hygrometer
Hygrometers Digital Thermo-Hygrometers
This digital thermo-hygrometer is designed for temperature and humidity measurements in fridges and freezers. It is suitable for use in food and beverage processing, storage and chemistries.
  • Switch the temperature readings from the internal sensor to the external probe
  • Maximum/minimum values, memory switchable and daily reset function
  • Large display shows all values simultaneously
  • The SET, ¹²/₂₄, ALERT function buttons are on the back of the unit, so the unit only has three buttons on the front
  • The display for the internal sensor is noted as 'INT' and the
More Product Information