VWR® Traceable®, Digital Thermometers
Supplier: VWR Collection
VWRI620-1826EA 121 SGD
VWRI620-1826
VWR® Traceable®, Digital Thermometers
Thermometers Digital Thermometers
These thermometers accurately monitor temperatures in freezers, refrigerators, incubators and environmental chambers. Bottle and solution insulate the sensor from transient temperature changes. Waterproof, non toxic glycol bottle probe.
  • Easy to read digital display
  • Maximum/minimum and current temperatures
W×D×H: 56×25×110 mm
Solution recognised as safe by the FDA.
Certifications: Solution recognised as safe by the FDA. Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with individually serialMore Product Information