VWR®, Digital Refrigerator and Freezer Thermometer
Supplier: VWR Collection
VWRI620-1913EA 40.3 SGD
VWRI620-1913
VWR®, Digital Refrigerator and Freezer Thermometer
Thermometers Digital Thermometers
Digital thermometer for refrigerators and freezers.
  • High and low alarm setting
  • Waterproof fridge sensor
  • °C/°F exchange
Calibration points: 0 and 50 °C
Certifications: This thermometer comes with a Traceable certificate.
Delivery information: Supplied with sensor holder, battery and VELCRO® magic tape.