VWR®, Digital Thermo-Hygrometer
Supplier: VWR Collection
VWRI620-1951EA 492 SGD
VWRI620-1951
VWR®, Digital Thermo-Hygrometer
Hygrometers Digital Thermo-Hygrometers
This handheld instrument is designed to measure temperature and humidity in air conditioning and ambient air monitoring. It provides a quick response in EDP rooms, galleries, offices, workshops, storage rooms, private buildings and greenhouses.
PT1000 and capacitive-polymer sensor (Lר: 35×14 mm).
Ordering information: Hygrometer is supplied with a test certificate and does not come with an ISO calibration certificate. If ISO calibration certificate is required, it has to specified during ordering. If calibration certificate is required after delivery, the unit has to be sent back forMore Product Information