VWR® Traceable® Digital Thermometer
Supplier: VWR Collection
VWRI620-1975EA 393 SGD
VWRI620-1975
VWR® Traceable® Digital Thermometer
Thermometers Digital Thermometers
Digital thermometer with extra long waterproof stainless steel 500 mm probe and 2,5 metre cable.
  • Large LCD readable at more than 20 metres away
  • Accurate readings with tip penetration of only 2,5 cm
  • Spring-powered holder enables probe to be placed anywhere
This thermometer is designed for placing in deep vessels like large beakers/flasks, vats, pails and drums in applications including foods, pharmaceuticals and industrial plants.
Certifications: Traceable to NIST. Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.