VWR® Traceable®, Wide Range Digital Thermometer
Supplier: VWR Collection
VWRI620-2004EA 724 SGD
VWRI620-2004
VWR® Traceable®, Wide Range Digital Thermometer
Thermometers Digital Thermometers
For quality control, routine monitoring, measuring temperatures in cuvettes, gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, petroleum products, foods and wastewater. Instant memory key recalls minimum and maximum readings over any time period. Stainless steel K-type thermocouple probe for any J-, K- or T-type probe.
  • Computer output allows thermometer to be connected to a computer or data logger for capturing and storing readings
  • Large, backlit LCD: Values can be read even in poor light
  • Count-up timer provides a relative
More Product Information