VWR® Traceable®, Digital Thermometer with Recorder Output
Supplier: VWR Collection
VWRI620-2046EA 335 SGD
VWRI620-2046
VWR® Traceable®, Digital Thermometer with Recorder Output
Thermometers Digital Thermometers
Type-K beaded probe with 120 mm cable
  • Recorder output provides a hard copy for today’s stringent QC documentation requirements
  • Maximum button captures the highest reading achieved
  • 'HOLD' button freezes display to capture current reading
  • Large, bright 4-digit LCD displays °F or °C values
  • Completely portable and designed for years of dependable and reliable service
This thermometer monitors conditions in chambers, clean rooms, food processing, scientific labs, and all temperature-sensitive manufacturing operations. It features a recorder jack thatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable