VWR® Traceable®, Digital Thermometer, Two Channels
Supplier: VWR Collection
VWRI620-2047EA 359 SGD
VWRI620-2047
VWR® Traceable®, Digital Thermometer, Two Channels
Thermometers Digital Thermometers
This thermometer provides recessed front panel adjustable offsets on two channels allowing the user to adjust the thermometer exactly to a specific temperature, in-house calibration, or particular sensor for increased accuracy. The rubberised case makes it a virtually environment-proof instrument for all weather, shock, and abusive conditions.
  • Large LCD shows temperatures of probe 1, probe 2, or the difference (delta) between them
  • Readout in °F or °C
  • 'MAX' button captures the highest reading achieved
  • 'HOLD' button freezes display to capture current reading
More Product Information