VWR® Traceable®, Digital Mini Thermometer
Supplier: VWR Collection
VWRI620-2055EA 85.9 SGD
VWRI620-2055
VWR® Traceable®, Digital Mini Thermometer
Thermometers Digital Thermometers
This small, powerful thermometer displays readings in °F and °C. The probe can be used to measure temperatures in liquid, air/gas or semi-solids. External stainless steel probe (Lר: 178×3,5 mm) with 45 cm cable.
  • Stainless steel probe is resistant to most chemicals
  • Business card size for carrying it with you: Fits into a toolkit, briefcase or coat pocket
  • Three-switch operation: HOLD function, On/Off button and switch between °C/°F
Delivery information: Supplied with a Traceable calibration certificate and 1 year silver-oxide battery.
Order Now
SPECIFICATIONS
Environmentally Preferable