VWR® Traceable® Full Scale (Plus) Digital Thermometers
Supplier: VWR Collection
VWRI620-2089EA 179 SGD
VWRI620-2089 VWRI620-2058 VWRI620-2064 VWRI620-1664
VWR® Traceable® Full Scale (Plus) Digital Thermometers
Thermometers Digital Thermometers
These thermometers, with LCD and magnetic back, enable placement on a metallic surface (incubators, freezers, water baths). They incorporate a flip-open easel for use on a lab bench, and a spring fastener for clipping to any edge.
  • Piercing tip
  • Feature available on 620-1664 and 620-2089 models: Reading updated every second
  • Model 620-2089: High/low alarm, maximum/minimum memories and 'HOLD' function
Stainless steel probe (Ø 3,5 mm) and 3 metre cable.
Certifications: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable