VWR® Traceable®, Shockproof Thermometer
Supplier: VWR Collection
VWRI620-2065EA 145 SGD
VWRI620-2065
VWR® Traceable®, Shockproof Thermometer
Thermometers Digital Thermometers
This 'flip to open' thermometer has a probe that can be positioned at any angle.
  • Easy to read large display with big digits
  • 'HOLD' function
  • Maximum/minimum memory readings over any time period
  • Folded up, the probe is protected allowing the thermometer to be carried on a wrist strap or in a pocket
Certifications: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with battery and Traceable® certificate of calibration.