VWR® Traceable®, Pocket Thermometer
Supplier: VWR Collection
VWRI620-2077EA 151 SGD
VWRI620-2077 VWRI620-2025
VWR® Traceable®, Pocket Thermometer
Thermometers Digital Thermometers
Safe thermometer for soil, cuvettes, test tubes, food products, incubators, petroleum hydrocarbons and virtually all lab tests. Readings are updated every second.
  • Unique, flat profile design, lightweight and built-in pocket clip to carry in a shirt pocket
  • 'HOLD' button freezes a reading for later observation. Switch from °F to °C
  • Memory function recalls maximum/minimum readings over any time period
Stainless steel probe (Ø×L: 3,5×89 mm).
Traceability to standards provided by NIST. Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
More Product Information