VWR® Traceable®, Waterproof Thermometer
Supplier: VWR Collection
VWRI620-2091EA 65.3 SGD
VWRI620-2091
VWR® Traceable®, Waterproof Thermometer
Thermometers Digital Thermometers
This piercing thermometer can measure the internal temperature of stored, frozen, or prepared food. For use in soil, brewing, food processing, refrigeration and pharmaceutical applications.
  • Waterproof construction allows it to be used in labs, plants, wash-down areas and in the field
  • Manufactured to be drop-proof from 1,5 metres
  • Reading (°C/°F) updated every second
Certifications: Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with probe, silver-oxide battery and Traceable® certificate of calibration.
Order Now
SPECIFICATIONS
Environmentally Preferable