VWR® Traceable®, Digital Thermometer with Data Log Memory
Supplier: VWR Collection
VWRI620-2094EA 117 SGD
VWRI620-2094
VWR® Traceable®, Digital Thermometer with Data Log Memory
Thermometers Digital Thermometers
This waterproof thermometer captures 10 temperature readings, stores them in memory, and recalls them on command. For food, agricultural, petrochemical and pharmaceutical applications. The waterproof construction allows use in wet labs, wash-down areas and outside.
  • Maximum/minimum memory readings over any time period
  • 'HOLD' function freezes the display to capture temperature to memory
  • Both high and low audible alarm may be programmed in 1,0 °C increments
  • Stores and recalls 10 temperature readings
Stainless steel probe (L×Ø: 190,5×6,4 mm) with 81,3 cmMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable