VWR® Traceable®, Digital Snap-In Thermometer
Supplier: VWR Collection
VWRI620-2097EA 66.3 SGD
VWRI620-2097
VWR® Traceable®, Digital Snap-In Thermometer
Thermometers Digital Thermometers
Digital thermometer with stainless steel probe and handle with 3 metre cable. Versatile panel-mount module snaps into place or can be used as a stand-alone unit.
  • 4-digit LCD: Easy to read
  • Maximum/minimum temperature memory
  • Readings available in °F or °C, updates every second
Delivery information: Supplied with Traceable certificate of calibration, probe, cable and battery.
Order Now
SPECIFICATIONS
Environmentally Preferable