VWR®, Bi-Metallic Dial Thermometer, Refrigerator/Freezer
Supplier: VWR Collection
VWRI620-2146EA 39.9 SGD
VWRI620-2146
VWR®, Bi-Metallic Dial Thermometer, Refrigerator/Freezer
Thermometers Analogue Thermometers
This thermometer, with analogue dial, indicates freezer, refrigerator, or spoilage temperatures.
  • Stainless steel model stands sturdily on rack shelves
  • Dual scale (°F/°C)
  • Red arrow for easier reading
  • Round, black modern design