Calibrated electronic verification thermometers, frio temp
Supplier: VWR Collection
VWRI620-0904EA 224 SGD
VWRI620-0904 VWRI620-2148 VWRI620-2344
Calibrated electronic verification thermometers, frio temp
Thermometers Digital Thermometers
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample.
  • Individually serialised
Plastic bottles cannot be used in ovens.
Certifications: ISO 17025, ISO/IEC 17025:2005, traceable to NIST.
Delivery information: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish), AAA or LR44 (620-2148) batteries.
Caution: * Internal
** External