VWR® Traceable®, Thermometers for Refrigerators and Freezers
Supplier: VWR Collection
89094-750
VWRI620-2211EA 404 SGD
VWRI620-2211 VWRI620-2212 VWRI620-2213 VWRI620-2214 VWRI620-2215 VWRI620-2216 VWRI620-2217 VWRU89094-750
VWR® Traceable®, Thermometers for Refrigerators and Freezers
Thermometers Digital Thermometers
These digital thermometers are designed for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Bottle probes are used in monitoring storage conditions for vaccines or other important samples.
  • Fixed 1 minute logging interval
  • Stores up to 10 unique alarm events
  • Capable of storing over one million observations
  • Raw data on unit cannot be overwritten or modified
  • Status: Low battery, low memory, USB data transfer and active alarm state
Bottle probe in non toxic glycol/bullet sensor/stainless steel probe with 3More Product Information