VWR®, Clock with Temperature and Humidity Display
Supplier: VWR Collection
VWRI620-2220EA 76.3 SGD
VWRI620-2220
VWR®, Clock with Temperature and Humidity Display
Timers Wall Clocks
This lab wall clock displays time, temperature, and humidity.
  • Quartz crystal accuracy
  • Analogue time, temperature (–20 to +55 °C) and humidity (0 to 100%)
  • Round modern design with highly visible blue colour
  • Bright red sweeping second hand provides superior visibility
Certifications: Traceability to standards provided by NIST. Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
Delivery information: Supplied with battery, wall-mount screw and certificate of calibration.