VWR® Radio-Controlled Wall Clock

Supplier: VWR Collection
VWRI620-2300EA 153 SGD
VWRI620-2300 VWRI620-2301
VWR® Radio-Controlled Wall Clock
Timers Wall Clocks
View time, temperature, and humidity from anywhere.

  • Calendar with month, date and day
  • Clock format of ¹²/₂₄ hours
  • Radio signal from United Kingdom (Anthorn, Cambria) or continental Europe (Mainflingen, Germany) atomic clock automatically sets correct date/time; calendar displays month/date, day, leap year, alarm
  • Giant-Digits™ 63 mm viewable from 30 m

W×D×H: 229×32×127 mm

Certifications: Traceability to standards provided by NIST. Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.

Delivery information: Supplied with batteries,More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR