VWR® Traceable®, Digital Refrigerator Thermometers
Supplier: VWR Collection
VWRI620-2302EA 46.5 SGD
VWRI620-2302
VWR® Traceable®, Digital Refrigerator Thermometers
Thermometers Digital Thermometers
This thermometer is designed for monitoring temperatures in storage rooms, labs, office and fume hoods. It includes attachment devices for flexible positioning.
  • Easy to read LCD
  • Switch from °F to °C
  • Updates every 30 seconds
Certifications: Traceability provided from ISO/IEC 17025 and standards by NIST.
Delivery information: Supplied with suction cup, VELCRO® fastener, silver-oxide battery and Traceable® certificate of calibration.
Order Now
SPECIFICATIONS
Environmentally Preferable