VWR® Traceable® Ultra, Triple-Display Thermometers
Supplier: VWR Collection
VWRI620-2334EA 168 SGD
VWRI620-2334 VWRI620-2335 VWRI620-2336 VWRI620-2337 VWRI620-2338 VWRI620-2339 VWRI620-2340
VWR® Traceable® Ultra, Triple-Display Thermometers
Thermometers Digital Thermometers
Digital thermometers are designed to accurately monitor the temperature of fridges, freezers, incubators, water baths or during transport, in one or two different areas with enclosed temperature-buffered sensors or bullet sensors.
  • Smart alarm: Visual and audible alarm even if the unit returns to non alarm conditions
  • Triple-display simultaneously shows maximum/minimum and current temperatures
  • Maximum/minimum indicator
Probes: 1 or 2 bullet sensors/1 or 2 bottle probes with non toxic glycol, in 2 versions; 5 ml bottle or micro bottle to simulateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable