VWR® Traceable®, Digital Thermometer, High Accuracy, Refrigerator and Freezer
Supplier: VWR Collection
VWRI620-2425EA 301 SGD
VWRI620-2425
VWR® Traceable®, Digital Thermometer, High Accuracy, Refrigerator and Freezer
Thermometers Digital Thermometers
This high accuracy thermometer monitors both ambient air and the temperature inside a refrigerator or freezer. It reads to 0,01 °C while monitoring MIN/MAX temperatures in the laboratory, and in the refrigerator or freezer simultaneously, with the exact date and time that the readings occurred. It can also be used in water baths, heating blocks and incubators.
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is
More Product Information