VWR® Calibratable Ultra-Accurate Thermometers
Supplier: VWR Collection
VWRI620-1932EA 840 SGD
VWRI620-1932 VWRI620-2652 VWRI620-2664
VWR® Calibratable Ultra-Accurate Thermometers
Thermometers Digital Thermometers
These ready to use thermometers, with probe, are designed for temperature measurements which require high accuracy. They can be used for general purpose applications in air, gas, liquid and solids measurements, depending on the probe type. They are suitable for use in pharmaceutical and biotechnology companies (production and laboratories in the food industry), dairy products, fish, meat, healthcare, blood, government institutes and the environment.
  • Low battery warning indicator
  • Calibratable: Offset and slope input
  • Maximum/minimum memory and 'HOLD'
More Product Information