VWR® TD23, Precise PT1000 Universal Waterproof Thermometer
Supplier: VWR Collection
VWRI620-2675EA 232 SGD
VWRI620-2675
VWR® TD23, Precise PT1000 Universal Waterproof Thermometer
Thermometers Digital Thermometers
Precise universal thermometer, with BNC connection, without sensor. Pure measurement with a focus on precision, speed and reliability packaged in a compact housing.This new handheld measuring device also impress with its ergonomic design, dust and water-protected design in accordance with IP 65/67 and the illuminated display. The compact thermometer is available with a practical BNC connection for interchangeable sensors.
  • Modern and functional housing
  • 3-line display/overhead display at the
  • Push of a button
  • Backlighting
  • Alarm function
  • Waterproof (IP 65/IP 67
More Product Information