VWR® TraceableLive®, CO₂, Humidity and Temperature Data Logger
Supplier: VWR Collection
VWRI620-2694EA 1660 SGD
VWRI620-2694
VWR® TraceableLive®, CO₂, Humidity and Temperature Data Logger
Data Loggers
TraceableLIVE® provides simple, digital monitoring of CO₂, temperature and humidity in greenhouses, micro labs, biofuel research and other environments. One free user license in standard delivery.
  • Ambient unit with dongle - place entire unit inside a greenhouse or other environments
  • Cable probe is ideal for CO₂ incubators
  • Alerts for CO₂, temperature, humidity, connectivity interruption and low battery
  • Secure SSL-encrypted data transmission
  • Alarm notifications available via Mobile Push, E-mail and SMS text message
  • Unlimited scalability, multiple point
More Product Information