VWR® Traceable® Kangaroo™, Digital Thermometer
Supplier: VWR Collection
VWRI620-2714EA 89 SGD
VWRI620-2714
VWR® Traceable® Kangaroo™, Digital Thermometer
Thermometers Digital Thermometers
Pocket-size thermometer features handy cable storage and probe brackets for portability.
  • Precision memory feature automatically captures and recalls minimum/maximum temperature readings over any time period, °F/°C switchable
  • Efficient alarm continues to sound every minute until temperature returns to non-alarm conditions
Traceable® Kangaroo™ thermometer carries the 106,7 cm (42") probe-cable in a reel-compartment pouch in the back of the unit. Perfect carry-around unit fits in any pocket, or use the convenient flip-open stand. Supplied silver-oxideMore Product Information