VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 122 mm Probe
Supplier: VWR Collection
VWRI620-2813EA 89.5 SGD
VWRI620-2813
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 122 mm Probe
Thermometers Digital Thermometers
Surface probe stem thermometer is ideal for cooking, laboratories and hot plates.
  • Stainless-steel 120 mm (4,7") probe keep hand away from hot surfaces
  • Will read temperatures in liquids, semi-solids and gases
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Certifications: Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: More Product Information