VWR® Traceable® Refrigerator/Freezer Digital Thermometers
Supplier: VWR Collection
VWRI620-2838EA 263 SGD
VWRI620-2838 VWRI620-2839 VWRI620-2840
VWR® Traceable® Refrigerator/Freezer Digital Thermometers
Thermometers Digital Thermometers
Reliably monitor and be alerted to any issues affecting the contents of your fridge or freezer without even opening the door.
  • Alarm indicator lights up brightly to draw your attention to an immediate issue
  • Know about excursions before they happen with the pre-alert alarm light that glows orange when your fridge or freezer is within 1 degree of your alarm setting
  • Use the mute button to silence the buzzer alarm while you are investigating the issue
  • Staff onboarding is a breeze with an enhanced, more intuitive user experience including large buttons, high-
More Product Information