VWR®, Temperature Indicating Strips
Supplier: VWR Collection
VWRI620-9102EA 29.3 SGD
VWRI620-9102 VWRI620-9103 VWRI620-9104 VWRI620-9105 VWRI620-9106
VWR®, Temperature Indicating Strips
Temperature Indicating Strips
These temperature indicating strips are designed for rapid measurements where conventional thermometers are impractical. They are suitable for use with equipment such as heating chambers and products which are sensitive at high temperatures.
  • Fast response within 1 second
  • Self-adhesive and irreversible - six points
  • Storage at maximum 25 °C for up to 2 years