VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWRI612-1681EA 46.6 SGD
VWRI612-1681 VWRI612-1747 VWRI612-1682 VWRI612-1683 VWRI612-1684 VWRI612-1685 VWRI612-1686 VWRI612-1687 VWRI612-1688 VWRI612-1756 VWRI612-1689 VWRI612-1690 VWRI612-2841 VWRI612-2842 VWRI612-2843 VWRI612-2844 VWRI612-2845 VWRI612-2846 VWRI612-2847 VWRI612-2848 VWRI612-2849 VWRI612-2850 VWRI612-2851 VWRI621-0058
VWR®, Transfer Pipettes, High Performance
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
  • Non toxic and inert to biological fluids and most acids
  • Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
  • Available sterile or non sterile
  • Sterility assurance level is SAL 10⁻⁶
Packaging: Sterile pipettes are packedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable