VWR®, Printer for VWR® Balances
Supplier: VWR Collection
VWRI630-1485EA 652 SGD
VWRI630-1485
VWR®, Printer for VWR® Balances
Printers Document Printers
A compact line thermal dot printer which can be used with VWR® balances.
  • Includes an internal clock
  • Prints date and time together with weight values
  • Print speed 50 mm/s
Delivery information: Supplied with connection cable.