VWR®, Immersion Oil
Supplier: VWR Collection
VWRI630-1808EA 41.7 SGD
VWRI630-1808
VWR®, Immersion Oil
Stains Microscopy and staining reagents
Immersion oil is used to increase microscope resolution.
  • For immersing of the lens and specimen
  • Transparent oil with high refractive index
  • Increases the numerical aperture of an objective