VWR® VisiScope® TL384 POL, Polarising Microscope
Supplier: VWR Collection
VWRI630-1936EA 5600 SGD
VWRI630-1936
VWR® VisiScope® TL384 POL, Polarising Microscope
Microscopes
Upright microscope for brightfield and polarising light observations with strain-free IOS N-plan POL objectives. Perfect for accurate polarisation analysis in biology and materials science.
  • Transmitted LED illumination for bright and clear images
  • Rotatable stage with locking mechanism
  • Rotatable analyser from 0° to 90° with graduated scale
  • Bertrand lens for conoscopic observation
  • All objectives with anti-fungus treatment
  • Coaxial coarse (adjustable tension) and fine focusing mechanism with limit stop to prevent contact between objective and specimen