VWR® VisiCam® TC 20 Plus, Camera with Tablet, Wireless
Supplier: VWR Collection
VWRI630-2661EA 1830 SGD
VWRI630-2661
VWR® VisiCam® TC 20 Plus, Camera with Tablet, Wireless
Cameras
This system is composed of a tablet and a wireless 5 MP camera. It can be attached to all microscopes with a C-mount.
  • No need for a computer
  • Wireless connection between tablet and camera: Tablet can be used in any position
  • Images can be shared immediately through wireless network
  • Tablet with user friendly 10,1" touch screen display
  • Quick and easy creation of data reports
Image analysis software pre-installed on the tablet has been developed so that user experience is easy and intuitive. The software allows preview, image capture, video recording,More Product Information