VWR® VisiScope® BL254 T1, Digital Microscope
Supplier: VWR Collection
VWRI630-2673EA 3510 SGD
VWRI630-2673
VWR® VisiScope® BL254 T1, Digital Microscope
Microscopes
Educational and laboratory microscope with integrated camera and removable tablet.
  • Die-cast frame for high stability and ergonomics during transmitted light observations
  • Tablet on the microscope rotatable in all directions and easily detached
  • Large opening behind the revolving nosepiece can be used as a handle
  • N-plan objectives with anti-fungus treatment
  • White illumination with light intensity controlled via a knob
  • Efficient LED energy saving lamp
Multilingual tablet with touch screen for wireless or Bluetooth® connection and powered by a two-cell Li-More Product Information