VWR® VisiScope® STB150, Stereo Microscope
Supplier: VWR Collection
VWRI630-2942EA 659 SGD
VWRI630-2942
VWR® VisiScope® STB150, Stereo Microscope
Microscopes
Binocular stereo microscope equipped with turnable objective (2× to 4×), fixed arm with handle, and bright, uniform LED illumination (transmitted and incident). Designed for educational purposes.
  • Adjustable dioptric compensation on both eyepiece tubes
  • Interpupillary distance adjustable between 53 and 75 mm
  • NiMH rechargeable battery for up to 8 hours of use
  • Efficient LED energy saving lamp
Ordering information: Rechargeable batteries not included.
Delivery information: Supplied ready to use with eyepieces, objective and dust cover.