VWR® VisiCam® B5, Digital USB Camera
Supplier: VWR Collection
VWRI630-2957EA 808 SGD
VWRI630-2957
VWR® VisiCam® B5, Digital USB Camera
Cameras
VisiCam B5 is a C-mount and eyepiece microscope camera.
  • C-mount connection for use on trinocular ports
  • Intuitive, flexible and easy to use
Recommended for educational and routine applications.
Delivery information: Supplied with software, calibration slide, USB cable (1,8 m), and eyepiece adapters for binocular and monocular microscopes (30 and 30,5 mm diameter).