VWR® VisiScope® SZT360-6 Stereo Microscope
Supplier: VWR Collection
VWRI630-2982EA 3010 SGD
VWRI630-2982
VWR® VisiScope® SZT360-6 Stereo Microscope
Microscopes
This compact Greenough stereo microscope with 6.72:1 zoom and 110 mm working distance is ideal for routine inspections in professional environments and laboratories.
  • Plan eyepieces with 23 mm field of view, high eye-point type, also suitable for use with spectacles
  • Double-adjustable incident and transmitted light (both LED) with separated brightness controls and 6300 K colour temperature
  • Specimen stage is fitted with a diffusing disc for transmitted light, specimen clips and a black/white disc
  • Pillar stand and ultra-flat base
  • Rack and pinion focusing
More Product Information