VWR® VisiScope® TL384B FL1 B, Fluorescence Microscope
Supplier: VWR Collection
VWRI630-3242EA 4770 SGD
VWRI630-3242
VWR® VisiScope® TL384B FL1 B, Fluorescence Microscope
Microscopes
These fluorescence microscopes are designed for routine lab applications.
  • Observation methods: Brightfield and LED fluorescence
  • Energy saving LED technology with higher light intensity for bright images and improved colour fidelity
  • Automatic light control and adjustment
  • Rackless stage with belt-driven mechanism allows a smooth movement
  • Five objectives with anti-fungus treatment
This extremely powerful LED fluorescence illuminator is combined with blue excitation filter set for the visualisation of the following fluorochromes: Acridine Yellow, AcridineMore Product Information