VWR® C-WF, Digital Camera
Supplier: VWR Collection
VWRI630-3271EA 860 SGD
VWRI630-3271
VWR® C-WF, Digital Camera
Cameras
Wi-Fi camera with good resolution (up to 5 MP), CMOS sensor and Wi-Fi connection, recommended for basic/general applications in education and home use. Work in a fully interactive, flexible and dynamic digital classroom and maximize the efficiency of learning processes.
  • Ideal for interactive, flexible and dynamic learning processes
  • Simple, intuitive operation with powerful software
  • Universal connection to any microscope brand
  • Direct eyepiece and trino port connection
  • Up to 5 MP images
  • Wi-Fi interface
  • Direct Wi-Fi connection (no router is required)
  • Image
More Product Information