VWR® Polysine®, Adhesion Slides
Supplier: VWR Collection
VWRI631-0107EA 62.2 SGD
VWRI631-0107
VWR® Polysine®, Adhesion Slides
Slides Adhesive Slides
These slides are coated with a permanent adhesive that provides excellent cell and tissue adhesion.
  • Tissue section thickness of 2 to 5 µm
  • 90° ground edges and colour-frosted end for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides
Recommended for molecular biology techniques and cytocentrifuged preparations to reduce tissue loss during prolonged DNA hybridisation and immunocytochemistry.
Order Now
SPECIFICATIONS
Environmentally Preferable