VWR®, Slide Folder
Supplier: VWR Collection
VWRI631-0689EA 13 SGD
VWRI631-0689 VWRI631-0690
VWR®, Slide Folder
Slide Holders
Slide trays hold 20 or 30 standard microscopy slides (76×26 mm).
  • Thumb-cut for easy slide removal
  • Horizontal compartments are recessed, protecting the specimens when the cover is closed
Colour: White (black rims)