VWR® Pipette Stands
Supplier: VWR Collection
VWRI631-0723EA 84.1 SGD
VWRI631-0723 VWRI631-0724 VWRI631-0725
VWR® Pipette Stands
Pipette Stands
Pipette stands made of transparent acrylic.
  • Three, four or six-place racks, accommodate most micropipettes
  • Width of upper notches: 25 mm
  • Width of lower notches: 20 mm
Order Now
SPECIFICATIONS
Environmentally Preferable