VWR®, Slide Holder Cabinet
Supplier: VWR Collection
VWRI631-0823EA 34 SGD
VWRI631-0823
VWR®, Slide Holder Cabinet
Slide Holders
Cardboard microscope slide holders offer instant reference and fast, fingertip retrieval.
  • Cardboard fitted with a protective plastic frame, which provides secure protection of specimens
  • Each slide holder card accepts four slides of any thickness
  • Space for identification and comments
Cards can be added, removed or changed for fast updating.