VWR® Q Path® Slide Organisers
Supplier: VWR Collection
VWRI631-1049EA 1510 SGD
VWRI631-1049
VWR® Q Path® Slide Organisers
Slide Holders
This metallic cabinet is suitable for the storage of 50×50 mm or 80×57 mm microscopy slides.
Weight: 9 kg
Weight: 9 kg
Order Now
SPECIFICATIONS
Environmentally Preferable