VWR® Q Path® Storage, 8 Drawers
Supplier: VWR Collection
VWRI631-1050EA 2410 SGD
VWRI631-1050
VWR® Q Path® Storage, 8 Drawers
Boxes Storage Boxes
Storage organiser with 2×4 metal drawers for embedding cassettes.
  • For 2600 cassettes
  • Weight: 18 kg