VWR® Q Path® Filing Cabinet for Cassettes
Supplier: VWR Collection
VWRI631-1065EA 111 SGD
VWRI631-1065
VWR® Q Path® Filing Cabinet for Cassettes
Boxes Storage Boxes
Filing cabinet in white cardboard.
  • Supplied assembled
  • Capacity for 410 cassettes
Two drawers.