VWR® Q Path® Slide Holders
Supplier: VWR Collection
VWRI631-1074EA 641 SGD
VWRI631-1074 VWRI631-1075
VWR® Q Path® Slide Holders
Slide Holders
Slide holders designed for applications in clinical cytology.
  • Fast, easy clip closure
  • For two or three slides, depending on the model
  • Modern, user friendly design
Order Now
SPECIFICATIONS
Environmentally Preferable